Anti-WHIP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24731
89361-474EA 610.96 USD
89361-474
Anti-WHIP Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to WHIP

ELISA: 1/5000 - 1/20000. WB: 1/200. Detects a band of approximately 97 kDa (predicted molecular weight: 72 kDa). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: WHIP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR