Anti-HILS1 Mouse Monoclonal Antibody [clone: 0.T.60]

GTX24269
89361-440EA 341.64 USD
89361-440
Anti-HILS1 Mouse Monoclonal Antibody [clone: 0.T.60]
Mouse monoclonal [0.T.60] to Histone H1 - ChIP Grade

For ELISA: Use at an assay dependent dilution. For ICC: Use at a concentration of 5-20 ?g/ml. For WB: Use at a concentration of 0.5-2.0 ?g/ml. Not tested in other applications. Optimal dilutions/concentrations should be determined by the researcher.

Type: Primary
Antigen: HILS1
Clonality: Monoclonal
Clone: 0.T.60
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR