Anti-TRIM63 Goat Polyclonal Antibody

Supplier: GeneTex
GTX24125
89361-422EA 611.09 USD
89361-422
Anti-TRIM63 Goat Polyclonal Antibody
Antibodies
Goat polyclonal to MURF1

ELISA: peptide ELISA (antibody detection limit dilution >1:32000). Western blot: no signal obtained yet but low background observed in human heart extracts at up to 1.0?g/ml.

Type: Primary
Antigen: TRIM63
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR