Anti-GZMB Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX24059
89352-094EA 643.29 USD
89352-094
Anti-GZMB Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to Granzyme B

Immunohistochemistry: use at a dilution of 1/50 - 1/100 for 30mins at RT (ABC method). Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. For formalin fixed paraffin embedded sections, perform heat mediated antigen retrieval using 10mM citrate buffer pH 6.0 before commencing with the IHC staining protocol.

Type: Primary
Antigen: GZMB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR