Anti-CD79a Rabbit Polyclonal Antibody

GTX24056
89352-090EA 397.47 USD
89352-090
Anti-CD79a Rabbit Polyclonal Antibody
Rabbit polyclonal to CD79a

Immunohistochemistry: use at a dilution of 1/50 - 1/100 for 30mins at RT (ABC method). Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. For formalin fixed paraffin embedded sections, perform heat mediated antigen retrieval using 10mM citrate buffer pH 6.0 before commencing with the IHC staining protocol.

Type: Primary
Antigen: CD79a
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR