Anti-TAZ Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX23961
89361-406EA 614.89 USD
89361-406
Anti-TAZ Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal to TAZ

Western blot: use at a dilution of 1/500 - 1/1000. Not tested in other applications. Optimal dilutions / concentrations should be determined by the end user. By Western blot, this antibody detects a band of ~49 kDa, which corresponds to the predicted molecular weight of TAZ.

Type: Primary
Antigen: TAZ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR