Anti-RPE65 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103472
89350-014EA 582.76 USD
89350-014
Anti-RPE65 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to RPE65 (retinal pigment epithelium-specific protein 65kDa)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: RPE65
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR