Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Thymine Dimer Mouse Monoclonal Antibody

Supplier: Genetex
GTX10347
Non-radioactive labeling of DNA is typically based on the enzymatic incorporation of modified nucleotides, carrying a small chemical moiety such as biotin, digoxigenin or fluorescein. These tags are subsequently detected by specific reagents such as streptavidin or a specific antibody coupled to a signal-producing enzyme. Although very efficient and reliable, labeling by in vitro polymerization is time-consuming, expensive, and may require various post-label purification steps to remove an excess of unincorporated precursors. An alternative strategy for DNA labeling, is based on the UV-induced formation of cyclobutane thymine dimers. Several methods have been described for the detection of thymine dimers, which are based on chromato-graphic analysis, and on biochemical analysis with endonucleases specific for UV-irradiated DNA. In addition, methods utilizing antibodies specific for pyrimidine dimers and other UV-induced DNA lesions have evolved, which permit the study of the induction and repair of these lesions without the requirement of in vivo radiolabeling of DNA. Photoimmunodetection, is a rapid, reliable and low-cost supplement to existing methods for nonradioactive DNA labeling. It enables a sensitive and non-radioactive method for labeling, detection, and quantification of high molecular weight (HMW) DNA fragments. The method is based on the introduction of thymine dimers into DNA after separa-tion by pulse field gel electrophoresis (PFGE), followed by detection with thymine dimer specific antibodies. The method does not require any enzymatic or chemical manipulation of the DNA sample. Monoclonal anti-bodies reacting specifically with thymine dimer, facilitate investigations on the apoptotic process and the role of UV-induced pyrimidine dimers in the process of photocarcinogenesis.

Type: Primary
Antigen: Thymine Dimer
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:




 

ORDER

Expand All / Collapse All
Supplier No. VWR Catalog Number Unit Price Quantity
GTX10347 89350-012 Each (25µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol Thymine Dimer
Antigen Name Thymine Dimer
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Host Mouse
Isotype IgG1
Western Blot Yes
Immunogen Chemical / Small Molecule.
Purification Protein G purified
Storage Buffer Phosphate buffered saline pH 7.4 containing 15mM sodium azide
Storage Temperature Keep as concentrated solution. Store at 4ºC short term. For extended storage aliquot and store at -20ºC or below. Avoid freeze-thaw cycles.
Tested Applications Blocking, Dot Blot, Slot Blot

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty