Anti-Dengue Virus NS5 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103350
89349-972EA 604.55 USD
89349-972
Anti-Dengue Virus NS5 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Dengue Virus NS5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR