Anti-Dengue Virus NS1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103346
89349-966EA 582.76 USD
89349-966
Anti-Dengue Virus NS1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Dengue Virus NS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR