Anti-Dengue Virus CAPSID Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX103343
89349-964EA 597.31 USD
89349-964
Anti-Dengue Virus CAPSID Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: Dengue Virus CAPSID
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dengue virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR