Anti-HLA-G Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101149
89348-850EA 582.76 USD
89348-850
Anti-HLA-G Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to HLA-G (major histocompatibility complex, class I, G)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: HLA-G
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR