Anti-LF Mouse Monoclonal Antibody [clone: 1A1]

Supplier: GeneTex
GTX10108
89350-630EA 696.18 USD
89350-630
Anti-LF Mouse Monoclonal Antibody [clone: 1A1]
Antibodies
Mouse monoclonal [1A1] to Lactoferrin

  • ELISA: Use at an assay dependent dilution. WB: Use at an assay dependent dilution. Detects a band of approximately 77 kDa. Can be used in an ELISA pair with GTX10110 (capture). Not tested in other applications. Optimal dilutions/concentrations should be determined by the end user.

Type: Primary
Antigen: LF
Clonality: Monoclonal
Clone: 1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR