Anti-PSAP Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX101065
89348-796EA 582.76 USD
89348-796
Anti-PSAP Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PSAP (prosaposin)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: PSAP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: 0
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR