Anti-CTSK Rabbit Polyclonal Antibody

GTX100898
89348-740EA 176.03 USD
89348-740
Anti-CTSK Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Cathepsin K (cathepsin K)

Antibody reactive against recombinant protein, detects immunogen. Further validation in progress.

Type: Primary
Antigen: CTSK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR