Anti-MYOD Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100885
89348-732EA 473.99 USD
89348-732
Anti-MYOD Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to MyoD (myogenic differentiation 1)

Suggested starting dilutions are as follows: WB: 1:100-1:1000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: MYOD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR