Anti-SLC27A4 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX100842
89348-710EA 582.76 USD
89348-710
Anti-SLC27A4 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to ACSVL4 (solute carrier family 27 (fatty acid transporter), member 4)

Suggested starting dilutions are as follows: WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Type: Primary
Antigen: SLC27A4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR