    Sort by:
  • Search Within Results
2-(1-Cyclohexenyl)ethylamine
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Supplier:  TCI America
Description:   CAS Number: 3399-73-3 MDL Number: MFCD00012058 Molecular Formula: C8H15N Molecular Weight: 125.22 Purity/Analysis Method: 98.0% (GC,T) Form: ...