    Sort by:
  • Search Within Results
1-(2-Chloroethyl)pyrrole
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
  List View Easy View
Sort by:
Catalog Number: (TCC1347-005G)
Supplier:  TCI America
Description:   CAS Number: 77200-24-9 MDL Number: MFCD00191330 Molecular Formula: C6H8ClN Molecular Weight: 129.59 Purity/Analysis Method: 98.0% (GC) Form: ...