    Sort by:
  • Search Within Results
2',4'-Dichlorobutyrophenone
SearchPresentationType-HORIZONTAL
Choose from the options below to refine your search. Multiple selections within any drop-down menu can be made. Click OK to update your results
SearchResultCount:"1"
Sort by:
Catalog Number: (TCD3468-25G)
Supplier:  TCI America
Description:   CAS Number: 66353-47-7 Molecular Formula: C10H10Cl2O Molecular Weight: 217.09 Purity/Analysis Method: 96.0% (GC) Form: Clear Liquid Boiling po...