Anti-CD16/CD32 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: FCR4G8]

NB100-63476
102112-608EA 536.92 USD
102112-608
Anti-CD16/CD32 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: FCR4G8]
The CD16 / CD32 Antibody (FCR4G8) [FITC] from Novus Biologicals is a rat monoclonal antibody to CD16 / CD32. This antibody reacts with mouse. The CD16 / CD32 Antibody (FCR4G8) [FITC] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: CD16/CD32
Clonality: Monoclonal
Clone: FCR4G8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR