Anti-CD16/CD32 Rat Monoclonal Antibody [clone: AP-MAB0815]

NBP1-06653
102157-392EA 493.85 USD
102157-392
Anti-CD16/CD32 Rat Monoclonal Antibody [clone: AP-MAB0815]
The CD16 / CD32 Antibody (AP-MAB0815) from Novus Biologicals is a rat monoclonal antibody to CD16 / CD32. This antibody reacts with mouse. The CD16 / CD32 Antibody (AP-MAB0815) has been validated for the following applications: Blocking / Neutralizing.

Type: Primary
Antigen: CD16/CD32
Clonality: Monoclonal
Clone: AP-MAB0815
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR