Anti-EMMPRIN/CD147 Rat Monoclonal Antibody [clone: OX114]

Supplier: Novus Biologicals
NB100-65518
102114-806EA 564.74 USD
102114-806
Anti-EMMPRIN/CD147 Rat Monoclonal Antibody [clone: OX114]
Antibodies
The EMMPRIN / CD147 Antibody (OX114) from Novus Biologicals is a rat monoclonal antibody to EMMPRIN / CD147. This antibody reacts with mouse. The EMMPRIN / CD147 Antibody (OX114) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.

Type: Primary
Antigen: EMMPRIN/CD147
Clonality: Monoclonal
Clone: OX114
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR