Anti-TNFSF9  Rat Monoclonal Antibody [clone: RM0344-6J43]

NBP2-11955
102250-646EA 493.85 USD
102250-646
Anti-TNFSF9  Rat Monoclonal Antibody [clone: RM0344-6J43]
The 4-1BB / TNFRSF9 / CD137 Antibody (RM0344-6J43) from Novus Biologicals is a rat monoclonal antibody to 4-1BB / TNFRSF9 / CD137. This antibody reacts with mouse. The 4-1BB / TNFRSF9 / CD137 Antibody (RM0344-6J43) has been validated for the following applications: Western Blot, Blocking / Neutralizing.

Type: Primary
Antigen: TNFSF9 
Clonality: Monoclonal
Clone: RM0344-6J43
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR