Anti-CD13 Rat Monoclonal Antibody [clone: ER-BMDM1]

Supplier: Novus Biologicals
NB100-64843
102114-206EA 491.08 USD
102114-206
Anti-CD13 Rat Monoclonal Antibody [clone: ER-BMDM1]
Antibodies
The Aminopeptidase N / CD13 Antibody (ER-BMDM1) from Novus Biologicals is a rat monoclonal antibody to Aminopeptidase N / CD13. This antibody reacts with mouse. The Aminopeptidase N / CD13 Antibody (ER-BMDM1) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Type: Primary
Antigen: CD13
Clonality: Monoclonal
Clone: ER-BMDM1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR