Anti-CD11b Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M1/70]

NBP1-27882
102161-252EA 253.85 USD
102161-252
Anti-CD11b Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: M1/70]
The CD11b Antibody (M1 / 70) [FITC] from Novus Biologicals is a rat monoclonal antibody to CD11b. This antibody reacts with mouse. The CD11b Antibody (M1 / 70) [FITC] has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Immunocytochemistry.

In vitro assay,

Type: Primary
Antigen: CD11b
Clonality: Monoclonal
Clone: M1/70
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR