Anti-CD11a Rat Monoclonal Antibody [clone: I21/7]

Supplier: Novus Biologicals
NBP1-27870
102161-232EA 564.74 USD
102161-232
Anti-CD11a Rat Monoclonal Antibody [clone: I21/7]
Antibodies
Rat Monoclonal CD11a Antibody (I21/7). Tested Applications: Flow Cytometry, Immunoblotting, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Mouse.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR