Anti-CD11a Rat Monoclonal Antibody [clone: I21/7] (Biotin)

Supplier: Novus Biologicals
NBP1-27872
102161-236EA 890.28 USD
102161-236
Anti-CD11a Rat Monoclonal Antibody [clone: I21/7] (Biotin)
Antibodies
Rat Monoclonal CD11a Antibody (I21/7) [Biotin]. Tested Applications: Flow Cytometry, Immunoblotting, Immunohistochemistry, Immunohistochemistry-Frozen, Immunoprecipitation. Tested Reactivity: Mouse.

Type: Primary
Antigen: CD11a
Clonality: Monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR