Anti-CD105 Rat Monoclonal Antibody [clone: MJ7/18]

Supplier: Novus Biologicals
NB100-77666
102117-242EA 417.23 USD
102117-242
Anti-CD105 Rat Monoclonal Antibody [clone: MJ7/18]
Antibodies
The Endoglin / CD105 Antibody (MJ7 / 18) from Novus Biologicals is a rat monoclonal antibody to Endoglin / CD105. This antibody reacts with mouse. The Endoglin / CD105 Antibody (MJ7 / 18) has been validated for the following applications: Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface).

Type: Primary
Antigen: CD105
Clonality: Monoclonal
Clone: MJ7/18
Conjugation: Unconjugated
Epitope: Endothelial cell selective TGB1 binding molecule-endoglin
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR