Anti-CCR6 Rat Monoclonal Antibody [clone: RM0187-25J7]

NBP2-12132
102251-000EA 493.85 USD
102251-000
Anti-CCR6 Rat Monoclonal Antibody [clone: RM0187-25J7]
The CCR6 Antibody (RM0187-25J7) from Novus Biologicals is a rat monoclonal antibody to CCR6. This antibody reacts with mouse. The CCR6 Antibody (RM0187-25J7) has been validated for the following applications: Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.

Type: Primary
Antigen: CCR6
Clonality: Monoclonal
Clone: RM0187-25J7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2 Alpha
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR