Anti-CCR3 Rat Monoclonal Antibody [clone: RM0186-36C4]

NBP2-12127
102250-990EA 493.85 USD
102250-990
Anti-CCR3 Rat Monoclonal Antibody [clone: RM0186-36C4]
The CCR3 Antibody (RM0186-36C4) from Novus Biologicals is a rat monoclonal antibody to CCR3. This antibody reacts with mouse. The CCR3 Antibody (RM0186-36C4) has been validated for the following applications: Flow Cytometry, CyTOF-ready.

Type: Primary
Antigen: CCR3
Clonality: Monoclonal
Clone: RM0186-36C4
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR