Anti-CCL14/HCC-1/HCC-3 Rat Monoclonal Antibody [clone: MM0138-4H14]

Supplier: Novus Biologicals
NBP2-12091
102250-918EA 379 USD
102250-918
Anti-CCL14/HCC-1/HCC-3 Rat Monoclonal Antibody [clone: MM0138-4H14]
Antibodies
The CCL14 / HCC-1 / HCC-3 Antibody (MM0138-4H14) from Novus Biologicals is a rat monoclonal antibody to CCL14 / HCC-1 / HCC-3. This antibody reacts with human. The CCL14 / HCC-1 / HCC-3 Antibody (MM0138-4H14) has been validated for the following applications: Western Blot, Blocking / Neutralizing.

Type: Primary
Antigen: CCL14/HCC-1/HCC-3
Clonality: Monoclonal
Clone: MM0138-4H14
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR