Anti-CCL12/MCP-5 Rat Monoclonal Antibody [clone: RM0169-8J25]

Supplier: Novus Biologicals
NBP2-12089
102250-914EA 635.51 USD
102250-914
Anti-CCL12/MCP-5 Rat Monoclonal Antibody [clone: RM0169-8J25]
Antibodies
The CCL12 / MCP-5 Antibody (RM0169-8J25) from Novus Biologicals is a rat monoclonal antibody to CCL12 / MCP-5. This antibody reacts with mouse. The CCL12 / MCP-5 Antibody (RM0169-8J25) has been validated for the following applications: Western Blot, Blocking / Neutralizing.

Type: Primary
Antigen: CCL12/MCP-5
Clonality: Monoclonal
Clone: RM0169-8J25
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR