Anti-Cathepsin L Rat Monoclonal Antibody [clone: RM0166-5G14]

NBP2-12082
102250-900EA 493.85 USD
102250-900
Anti-Cathepsin L Rat Monoclonal Antibody [clone: RM0166-5G14]
The Cathepsin L Antibody (RM0166-5G14) from Novus Biologicals is a rat monoclonal antibody to Cathepsin L. This antibody reacts with mouse. The Cathepsin L Antibody (RM0166-5G14) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: Cathepsin L
Clonality: Monoclonal
Clone: RM0166-5G14
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR