Anti-Cathepsin 3 Rat Monoclonal Antibody [clone: RM0156-2K13]

Supplier: Novus Biologicals
NBP2-12422
102251-506EA 635.51 USD
102251-506
Anti-Cathepsin 3 Rat Monoclonal Antibody [clone: RM0156-2K13]
Antibodies
The Cathepsin 3 Antibody (RM0156-2K13) from Novus Biologicals is a rat monoclonal antibody to Cathepsin 3. This antibody reacts with mouse. The Cathepsin 3 Antibody (RM0156-2K13) has been validated for the following applications: Western Blot.

Type: Primary
Antigen: Cathepsin 3
Clonality: Monoclonal
Clone: RM0156-2K13
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR