Anti-BCMA/TNFRSF17 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Vicky-1]

NBP1-97655
102222-238EA 564.62 USD
102222-238
Anti-BCMA/TNFRSF17 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Vicky-1]
The BCMA / TNFRSF17 Antibody (Vicky-1) [FITC] from Novus Biologicals is a rat monoclonal antibody to BCMA / TNFRSF17. This antibody reacts with human. The BCMA / TNFRSF17 Antibody (Vicky-1) [FITC] has been validated for the following applications: Flow Cytometry.

Type: Primary
Antigen: BCMA/TNFRSF17
Clonality: Monoclonal
Clone: Vicky-1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR