Anti-BCMA/TNFRSF17 Rat Monoclonal Antibody [clone: RM0336-17B1]

NBP2-11939
102250-614EA 493.85 USD
102250-614
Anti-BCMA/TNFRSF17 Rat Monoclonal Antibody [clone: RM0336-17B1]
The BCMA / TNFRSF17 Antibody (RM0336-17B1) from Novus Biologicals is a rat monoclonal antibody to BCMA / TNFRSF17. This antibody reacts with mouse. The BCMA / TNFRSF17 Antibody (RM0336-17B1) has been validated for the following applications: Western Blot.

Type: Primary
Antigen: BCMA/TNFRSF17
Clonality: Monoclonal
Clone: RM0336-17B1
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR