Anti-ART2.2/Art2b Rat Monoclonal Antibody [clone: Nika102]

Supplier: Novus Biologicals
NB100-65853
102115-044EA 564.74 USD
102115-044
Anti-ART2.2/Art2b Rat Monoclonal Antibody [clone: Nika102]
Antibodies
The ART2.2 / Art2b Antibody (Nika102) from Novus Biologicals is a rat monoclonal antibody to ART2.2 / Art2b. This antibody reacts with mouse. The ART2.2 / Art2b Antibody (Nika102) has been validated for the following applications: Flow Cytometry, Immunoprecipitation.

Type: Primary
Antigen: ART2.2/Art2b
Clonality: Monoclonal
Clone: Nika102
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR