Rabbit IgG Polyclonal Isotype Control (FITC (Fluorescein Isothiocyanate))

NBP1-43957
102173-706EA 253.85 USD
102173-706
Rabbit IgG Polyclonal Isotype Control (FITC (Fluorescein Isothiocyanate))
Rabbit Polyclonal Rabbit IgG Isotype Control [FITC]. Tested Applications: Flow Cytometry. Tested Reactivity: All Species.

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: pan
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR