Rabbit IgG Polyclonal Isotype Control (Agarose)

NB600-441
102133-982EA 395.38 USD
102133-982
Rabbit IgG Polyclonal Isotype Control (Agarose)
Rabbit Polyclonal Rabbit IgG Isotype Control [Agarose Immobilized]. Tested Applications: Immunoprecipitation. Tested Reactivity: Rabbit.

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Agarose immobilized
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR