Anti-IgG Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-73813
102204-646EA 267.51 USD
102204-646
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Rabbit anti-Turkey IgG, H/L Chains Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Turkey.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Turkey
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR