Anti-IgG Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-72744
102203-142EA 295.82 USD
102203-142
Anti-IgG Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal Rabbit anti-Sheep IgG, H/L chains Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Sheep.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR