Anti-IgG Rabbit Polyclonal Antibody (DyLight® 800)

Supplier: Novus Biologicals
NBP1-73000
102203-512EA 225.05 USD
102203-512
Anti-IgG Rabbit Polyclonal Antibody (DyLight® 800)
Antibodies
Rabbit Polyclonal Rabbit anti-Sheep IgG, H/L chains Antibody [DyLight 800]. Tested Applications: Western Blot, Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Sheep.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DyLight® 800
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR