Anti-IgG Rabbit Polyclonal Antibody (Cy5®)

NBP1-73022
102203-534EA 536.92 USD
102203-534
Anti-IgG Rabbit Polyclonal Antibody (Cy5®)
Rabbit Polyclonal Rabbit anti-Sheep IgG, H/L chains Antibody [Cy5]. Tested Applications: Fluorophore-linked immunosorbent assay, Immunocytochemistry/Immunofluorescence. Tested Reactivity: Sheep.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity: Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR