Anti-VSIG4 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-69631
102199-212EA 578.89 USD
102199-212
Anti-VSIG4 Rabbit Polyclonal Antibody
Antibodies
The VSIG4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to VSIG4. This antibody reacts with human. The VSIG4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: VSIG4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR