Anti-VPS37C Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-31068
102164-092EA 635.51 USD
102164-092
Anti-VPS37C Rabbit Polyclonal Antibody
Antibodies
The VPS37C Antibody from Novus Biologicals is a rabbit polyclonal antibody to VPS37C. This antibody reacts with human. The VPS37C Antibody has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence.

Type: Primary
Antigen: VPS37C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR