Anti-VPS26 Rabbit Polyclonal Antibody

NBP1-30909
102163-862EA 493.85 USD
102163-862
Anti-VPS26 Rabbit Polyclonal Antibody
The VPS26 Antibody from Novus Biologicals is a rabbit polyclonal antibody to VPS26. This antibody reacts with human. The VPS26 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: VPS26
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR