Anti-VPS11 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-04092
102234-898EA 635.51 USD
102234-898
Anti-VPS11 Rabbit Polyclonal Antibody
Antibodies
The VPS11 Antibody from Novus Biologicals is a rabbit polyclonal antibody to VPS11. This antibody reacts with human. The VPS11 Antibody has been validated for the following applications: Western Blot, Immunoprecipitation.

Type: Primary
Antigen: VPS11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR