Anti-Von Hippel Lindau Rabbit Polyclonal Antibody

NB100-1899SS
102106-310EA 112.31 USD
102106-310
Anti-Von Hippel Lindau Rabbit Polyclonal Antibody
The Von Hippel Lindau Antibody from Novus Biologicals is a rabbit polyclonal antibody to Von Hippel Lindau. This antibody reacts with human. The Von Hippel Lindau Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.

Recommended Dilutions: Immunohistochemistry (Parafin): 1:50-1:200

Type: Primary
Antigen: Von Hippel Lindau
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR