Anti-Vitamin D BP Rabbit Polyclonal Antibody

NB600-1504
102133-416EA 253.85 USD
102133-416
Anti-Vitamin D BP Rabbit Polyclonal Antibody
The Vitamin D BP Antibody from Novus Biologicals is a rabbit polyclonal antibody to Vitamin D BP. This antibody reacts with human. The Vitamin D BP Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: Vitamin D BP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR